Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TR2/NR2C1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB291186 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291186 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291186 Supplier Novus Biologicals Supplier No. NBP171803PCP

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

TR2/NR2C1 Polyclonal antibody specifically detects TR2/NR2C1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen TR2/NR2C1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation
Formulation PBS
Gene Alias nuclear receptor subfamily 2 group C member 1, nuclear receptor subfamily 2, group C isoform, nuclear receptor subfamily 2, group C, member 1, Orphan nuclear receptor TR2, Testicular receptor 2, TR2, TR2 nuclear hormone receptor, TR2-11
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of human Testicular Receptor 2 using the numbering given in entry NP_003288.2 (GeneID 7181).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7181
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.