Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TR4/NR2C2 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB101051 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB101051 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB101051 Supplier Novus Biologicals Supplier No. NBP297138V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TR4/NR2C2 Polyclonal specifically detects TR4/NR2C2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TR4/NR2C2
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias hTAK1, Nuclear hormone receptor TR4, nuclear receptor subfamily 2 group C member 2, nuclear receptor subfamily 2, group C, member 2, Orphan nuclear receptor TAK1, Orphan nuclear receptor TR4, TAK1TR4TR2R1, Testicular receptor 4, TR4 nuclear hormone receptor
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TR4/NR2C2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7182
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.