Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TR4/NR2C2 Antibody (OTI4B1), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP27477A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27477A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27477A70 Supplier Novus Biologicals Supplier No. NBP274577AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TR4/NR2C2 Monoclonal specifically detects TR4/NR2C2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TR4/NR2C2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4B1
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias hTAK1, Nuclear hormone receptor TR4, nuclear receptor subfamily 2 group C member 2, nuclear receptor subfamily 2, group C, member 2, Orphan nuclear receptor TAK1, Orphan nuclear receptor TR4, TAK1TR4TR2R1, Testicular receptor 4, TR4 nuclear hormone receptor
Gene Symbols NR2C2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-241 of human TR4/NR2C2 (NP_003289) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7182.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.