Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRA-1-85/CD147 Antibody (TRA-1-85) [DyLight 488], Novus Biologicals™
SDP

Catalog No. NB372830 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372830 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372830 Supplier Novus Biologicals Supplier No. FAB3195K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRA-1-85/CD147 Monoclonal specifically detects TRA-1-85/CD147 in Human samples. It is validated for CyTOF, Flow Cytometry, Gene Knock-Out, Immunocytochemistry, Western Blot.

Specifications

Antigen TRA-1-85/CD147
Applications CyTOF, Flow Cytometry, Gene Knock-Out, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone TRA-1-85
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen 2120Ep human embryonal carcinoma cell line
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.