Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRA2B Antibody, Novus Biologicals™
SDP

Catalog No. NBP157457 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP157457 100 μL
NBP15745720 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP157457 Supplier Novus Biologicals Supplier No. NBP157457
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRA2B Polyclonal specifically detects TRA2B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TRA2B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P62995
Gene Alias DKFZp686F18120, hTRA2-beta, SFRS10, Splicing factor, arginine/serine-rich 10, splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila), SRFS10, TRA-2 beta, TRA2-beta, TRAN2B, transformer 2 beta homolog (Drosophila), transformer 2 homolog, Transformer-2 protein homolog B, transformer-2 protein homolog beta, transformer-2-beta
Gene Symbols TRA2B
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SFRS10 (splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila)) The peptide sequence was selected from the middle region of SFRS10. Peptide sequence GVFGLSLYTTERDLREVFSKYGPIADVSIVYDQQSRRSRGFAFVYFENVD The peptide sequence for this immunogen was taken from within the described region.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6434
Test Specificity Expected identity based on immunogen sequence: Zebrafish: 92%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.