Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRA2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$501.50
Specifications
Antigen | TRA2B |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157457
![]() |
Novus Biologicals
NBP157457 |
100 μL |
Each for $501.50
|
|
|||||
NBP15745720
![]() |
Novus Biologicals
NBP15745720UL |
20 μL | N/A | N/A | N/A | ||||
Description
TRA2B Polyclonal specifically detects TRA2B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TRA2B | |
Polyclonal | |
Purified | |
RUO | |
DKFZp686F18120, hTRA2-beta, SFRS10, Splicing factor, arginine/serine-rich 10, splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila), SRFS10, TRA-2 beta, TRA2-beta, TRAN2B, transformer 2 beta homolog (Drosophila), transformer 2 homolog, Transformer-2 protein homolog B, transformer-2 protein homolog beta, transformer-2-beta | |
TRA2B | |
Primary |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
P62995 | |
6434 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title