Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trace Amine Receptor 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224720SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224720SS 0.025 mg
NBP224720 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224720SS Supplier Novus Biologicals Supplier No. NBP224720SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Trace Amine Receptor 1 Polyclonal specifically detects Trace Amine Receptor 1 in Human, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Trace Amine Receptor 1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q96RJ0
Gene Alias MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1
Gene Symbols TAAR1
Host Species Rabbit
Immunogen A portion of amino acids 225-250 of human TAAR1 was used as the immunogen
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 134864
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 86% homologous in mouse and 80% homologous in dog.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.