Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TrackIt™ Cyan/Orange Loading Buffer
GREENER_CHOICE

Catalog No. 10482028
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 0.5 mL
Catalog No. Quantity
10-482-028 3 x 0.5 mL
1 options

Catalog No. 10-482-028

Supplier: Invitrogen™ 10482028

Only null left
Add to Cart
Add to Cart

Includes

TrackIt™ Cyan/Orange Loading Buffer is composed of 30% (v/v) glycerol, 60mM Tris-HCl (pH 7.5), 60mM EDTA, 0.36% (w/v) XCFF, and 2.4% (w/v) Orange G. TrackIt™ Cyan/Yellow Loading Buffer is composed of 30% (v/v) glycerol, 60mM Tris-HCl (pH 7.5), 60mM EDTA, 0.36% (w/v) XCFF, and 3.6% (w/v) Tartrazine

A 6X solution used for easy loading and tracking of DNA samples in agarose gels

TrackIt™ Loading Buffers loading buffers contain two tracking dyes, one that runs behind sample and one that runs ahead of sample. The tracking dyes serve as visual markers for following migration during electrophoresis and indicate when maximum resolution of DNA fragments has been achieved. Additionally, visualization of DNA bands will not be obscured by tracking dyes because they run outside limits of most DNA samples.

 

  • TrackIt Cyan/Orange Loading Buffer is recommended for DNA fragments between 50 bp and 10 Kb
  • TrackIt Cyan/Yellow Loading Buffer is recommended for DNA fragments between 10 bp and 1 Kb
  • For use with agarose and E-Gel™ gels
  •  

    Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 3 x 0.5 mL 6X TrackIt Cyan/Orange Loading Buffer

Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.

Buffer Sample Loading Buffers
Concentration 6X
Gel Type Agarose
Label or Dye Orange G, Xylene Cyanol, Xylene Cyanol FF (XCFF)
Sample Loading Volume 0.5 mL
Gel Compatibility Agarose Gels
Quantity 3 x 0.5 mL
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Line TrackIt
Product Type Loading Buffer
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.