Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRACP/PAP/ACP5 Antibody (5C5E7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP237362 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237362 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237362 Supplier Novus Biologicals Supplier No. NBP237362
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRACP/PAP/ACP5 Monoclonal specifically detects TRACP/PAP/ACP5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen TRACP/PAP/ACP5
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 5C5E7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias HPAP, TRACP5a, TRACP5b, TRAP, TrATPase
Gene Symbols ACP5
Host Species Mouse
Immunogen Purified recombinant fragment of human ACP5 (AA: 221-325) expressed in E. Coli.
Molecular Weight of Antigen 36.6 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.