Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRACP/PAP/ACP5 Antibody (5C5E7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | TRACP/PAP/ACP5 |
---|---|
Clone | 5C5E7 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Description
TRACP/PAP/ACP5 Monoclonal specifically detects TRACP/PAP/ACP5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
TRACP/PAP/ACP5 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
RUO | |
PBS with 0.05% Sodium Azide | |
54 | |
Purified recombinant fragment of human ACP5 (AA: 221-325) expressed in E. Coli. | |
Primary | |
36.6 kDa |
5C5E7 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
Monoclonal | |
Mouse | |
Human | |
HPAP, TRACP5a, TRACP5b, TRAP, TrATPase | |
ACP5 | |
IgG1 | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title