Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRACP/PAP/ACP5 Antibody (ACP5/2336R), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB384991 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384991 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384991 Supplier Novus Biologicals Supplier No. NBP308700AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TRACP/PAP/ACP5 Monoclonal antibody specifically detects TRACP/PAP/ACP5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TRACP/PAP/ACP5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ACP5/2336R
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias HPAP, TRACP5a, TRACP5b, TRAP, TrATPase
Host Species Rabbit
Immunogen Recombinant full-length human TRACP/PAP/ACP5 protein (Uniprot: P13686)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 54
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.