Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRADD Antibody (313203), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB304329 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304329 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304329 Supplier Novus Biologicals Supplier No. FAB2658N

Mouse Monoclonal Antibody

TRADD Monoclonal antibody specifically detects TRADD in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen TRADD
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 313203
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias MGC11078, TNFRSF1A-associated via death domainTNFR1-associated DEATH domain protein, tumor necrosis factor receptor type 1 associated death domain protein, tumor necrosis factor receptor type 1-associated DEATH domain protein, tumor necrosis factor receptor-1-associated protein
Host Species Mouse
Immunogen E. coli-derived recombinant human TRADD, Met1-Ala312, Accession # Q15628
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Death Receptor Signaling Pathway, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8717
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.