Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB10056173
Description
TRAF-2 Polyclonal specifically detects TRAF-2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| TRAF-2 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| The gene encoding TRAF2 produces, by alternative splicing, 15 different transcripts, all with introns, putatively encoding 15 different protein isoforms. These isoforms are of varying amino acid lengths. Therefore the molecular weight observed by western blot may vary, depending on the isoform expressed. Users are encouraged to use the peptide immunogen sequence information provided in the antigen section to gain additional information about the potential TRAF2 isoforms detected by the antibody. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| TRAF2 | |
| A synthetic peptide corresponding to amino acids 6-31 (VTPPGSLELLQPGFSKTLLGTKLEAK) of human TRAF2 was used as immunogen, GenBank no. NP_066961.2. This peptide sequence is 92% conserved in mouse and rat TRAF2. The peptide immunogen sequence is 100% conserved in multiple human TRAF2 GenBank accession numbers. These TRAF2 accession numbers are of varying amino acid (aa) lengths and include: 1) NP_066961.2 = 501 aa. 2) CAI12705.1 =121 aa. 3) CAI12704.1 = 422 aa. 4) CAI15104.1 = 193 aa. 5) BAD96681.1 = 205 aa. | |
| 0.05 mL | |
| Apoptosis, Cancer, Cytokine Research, Zinc Finger | |
| 7186 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction