Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF-3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB357699 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357699 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357699 Supplier Novus Biologicals Supplier No. NBP176642MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRAF-3 Polyclonal antibody specifically detects TRAF-3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen TRAF-3
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CAP-1CD40 receptor associated factor 1, CD40 associated protein 1, CD40 binding protein, CD40 receptor-associated factor 1, CD40-binding protein, CD40bp, CRAF1CAP1, LAP1CD40BP, LMP1-associated protein 1, TNF receptor-associated factor 3
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human. The immunogen is located within amino acids 20 - 70 of TRAF3. Amino Acid Squence: GYKEKFVKTVEDKYK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology, Innate Immunity, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7187
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.