Learn More
Description
Specifications
Specifications
| Antigen | TRAF-3 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:4000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:4000 |
| Formulation | Whole antisera with 0.02% Sodium Azide |
| Gene Alias | CAP-1CD40 receptor associated factor 1, CD40 associated protein 1, CD40 binding protein, CD40 receptor-associated factor 1, CD40-binding protein, CD40bp, CRAF1CAP1, LAP1CD40BP, LMP1-associated protein 1, TNF receptor-associated factor 3 |
| Gene Symbols | TRAF3 |
| Host Species | Rabbit |
| Immunogen | A recombinant protein fragment corresponding to the cytoplasmic end of human TRAF3 (amino acids 341-568) was used as immunogen, GenBank no. GenBank no. gi 22027616 ref NP_003291.2 . |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
