Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF-4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

Supplier:  Novus Biologicals NB10056177

 View more versions of this product

Catalog No. NB10056177


Only null left
Add to Cart

Description

Description

TRAF-4 Polyclonal specifically detects TRAF-4 in Human, Mouse, Rat, Bovine, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

TRAF-4
Polyclonal
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:1000-1:5000, Immunohistochemistry-Paraffin 1:1000-1:5000
CART1RING finger protein 83, Cysteine-rich domain associated with RING and Traf domains protein 1, Metastatic lymph node gene 62 protein, MLN 62, MLN62cysteine-rich domain associated with ring and TRAF domain, RNF83malignant 62, TNF receptor-associated factor 4, tumor necrosis receptor-associated factor 4A
Rabbit
Unpurified
RUO
Primary
TRAF4
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Whole antisera with 0.02% Sodium Azide
TRAF4
A synthetic peptide corresponding to amino acids 1-17 (MPGFDYKFLEKPKRRLL) of human TRAF4 was used as immunogen; GenBank no. gi 22027622 ref NP_004286.2 .
0.05 mL
Cancer, Zinc Finger
9618
Human, Mouse, Rat, Bovine, Zebrafish
Antisera
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only