Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF-4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$206.50 - $316.50
Specifications
Antigen | TRAF-4 |
---|---|
Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:1000-1:5000, Immunohistochemistry-Paraffin 1:1000-1:5000 |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056177T
![]() |
Novus Biologicals
NB10056177SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB10056177
![]() |
Novus Biologicals
NB10056177 |
0.05 mL |
Each for $316.50
|
|
|||||
Description
TRAF-4 Polyclonal specifically detects TRAF-4 in Human, Mouse, Rat, Bovine, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
TRAF-4 | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Zinc Finger | |
Whole antisera with 0.02% Sodium Azide | |
9618 | |
A synthetic peptide corresponding to amino acids 1-17 (MPGFDYKFLEKPKRRLL) of human TRAF4 was used as immunogen; GenBank no. gi 22027622 ref NP_004286.2 . | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:1000-1:5000, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat, Bovine, Zebrafish | |
CART1RING finger protein 83, Cysteine-rich domain associated with RING and Traf domains protein 1, Metastatic lymph node gene 62 protein, MLN 62, MLN62cysteine-rich domain associated with ring and TRAF domain, RNF83malignant 62, TNF receptor-associated factor 4, tumor necrosis receptor-associated factor 4A | |
TRAF4 | |
IgG | |
Unpurified | |
TRAF4 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title