Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF-5 Antibody (55A219), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N601107A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N601107A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N601107A350 Supplier Novus Biologicals Supplier No. NB6001107AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAF-5 Monoclonal specifically detects TRAF-5 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRAF-5
Applications Western Blot
Classification Monoclonal
Clone 55A219
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias MGC:39780, RNF84RING finger protein 84, TNF receptor-associated factor 5
Gene Symbols TRAF5
Host Species Mouse
Immunogen Anti-TRAF5 monoclonal antibody was raised against a fusion protein corresponding to amino acids 77 to 186 of human TRAF5.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7188
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.