Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF-5 Antibody (55A219), Biotin, Novus Biologicals™
SDP

Catalog No. NB6001107B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001107B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001107B Supplier Novus Biologicals Supplier No. NB6001107B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAF-5 Monoclonal specifically detects TRAF-5 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRAF-5
Applications Western Blot
Classification Monoclonal
Clone 55A219
Conjugate Biotin
Dilution Western Blot
Gene Accession No. O00463
Gene Alias MGC:39780, RNF84RING finger protein 84, TNF receptor-associated factor 5
Gene Symbols TRAF5
Host Species Mouse
Immunogen Anti-TRAF5 monoclonal antibody was raised against a fusion protein corresponding to amino acids 77 to 186 of human TRAF5.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7188
Test Specificity Cross-reacts with Human. Not yet tested in other species.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.