Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF-6 Antibody (326019), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348441 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348441 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348441 Supplier Novus Biologicals Supplier No. FAB3284MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAF-6 Monoclonal specifically detects TRAF-6 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen TRAF-6
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 326019
Conjugate mFluor Violet 450 SE
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6
Host Species Mouse
Immunogen E.coli-derived recombinant human TRAF-6, Met1-Val522, Accession # Q9Y4K3
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7189
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.