Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF-6 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056436T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056436T 0.025 mg
NB10056436 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056436T Supplier Novus Biologicals Supplier No. NB10056436SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

TRAF-6 Polyclonal specifically detects TRAF-6 in Human samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen TRAF-6
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5 ug/ml, Flow Cytometry
Formulation 0.1 ml PBS with 0.02% Sodium Azide
Gene Accession No. Q9Y4K3
Gene Alias E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6
Gene Symbols TRAF6
Host Species Rabbit
Immunogen Anti-TRAF6 polyclonal antibody was raised against a peptide corresponding to amino acids between 410 and 460 of human TRAF6.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7189
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.