Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TRAF-6 Inhibitor Peptide Set
SDP

Applications: In vitro assay, Block/Neutralize

$706.00 - $1244.00

Specifications

Host Species Human, Mouse, Rat
Color White
Components TRAF6 Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKRKIPTEDEY (TRAF6 binding sequence: RKIPTEDEY). Molecular weight: 3494. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361.
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type TRAF-6 Inhibitor Peptide Set
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP226506
SDP
View Documents
Novus Biologicals™
NBP226506
2 mg
Each for $706.00
Only null left
Add to Cart
 
NBP2265065
SDP
View Documents
Novus Biologicals™
NBP2265065MG
5 mg
Each for $1,244.00
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

Human, Mouse, Rat
TRAF6 Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKRKIPTEDEY (TRAF6 binding sequence: RKIPTEDEY). Molecular weight: 3494. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361.
TRAF-6 Inhibitor Peptide Set
TRAF-6
White
Store at -20°C. Avoid freeze-thaw cycles.
3494
Lyophilized Solid
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.