Learn More
Description
Specifications
Specifications
| Antigen | TRAF3IP1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml |
| Formulation | PBS containing 0.05% BSA with 0.05% Sodium Azide |
| Gene Accession No. | Q8TDR0 |
| Gene Alias | DKFZp434F124, Interleukin-13 receptor alpha 1-binding protein 1, Microtubule-interacting protein associated with TRAF3, microtubule-interacting protein that associates with TRAF3, MIPT3interleukin 13 receptor alpha 1-binding protein-1, MIP-T3microtubule interacting protein that associates with TRAF3, TNF receptor-associated factor 3 interacting protein 1, TRAF3-interacting protein 1 |
| Gene Symbols | TRAF3IP1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
