Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF3IP1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB332252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332252 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332252 Supplier Novus Biologicals Supplier No. NBP297341PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRAF3IP1 Polyclonal antibody specifically detects TRAF3IP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TRAF3IP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DKFZp434F124, Interleukin-13 receptor alpha 1-binding protein 1, Microtubule-interacting protein associated with TRAF3, microtubule-interacting protein that associates with TRAF3, MIPT3interleukin 13 receptor alpha 1-binding protein-1, MIP-T3microtubule interacting protein that associates with TRAF3, TNF receptor-associated factor 3 interacting protein 1, TRAF3-interacting protein 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TRAF3IP1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26146
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.