Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF3IP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159996
Description
TRAF3IP3 Polyclonal specifically detects TRAF3IP3 in Human samples. It is validated for Western Blot.Specifications
TRAF3IP3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
DJ434O14.3, FLJ44151, MGC117354, MGC163289, T3JAMTRAF3-interacting protein 3, TRAF3 interacting Jun N terminal kinase (JNK) activating modulator, TRAF3 interacting protein 3, TRAF3-interacting JNK-activating modulator | |
Rabbit | |
61 kDa | |
100 μL | |
Primary | |
Guinea pig 86%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9Y228 | |
TRAF3IP3 | |
Synthetic peptides corresponding to TRAF3IP3(TRAF3 interacting protein 3) The peptide sequence was selected from the C terminal of TRAF3IP3. Peptide sequence DLQDQLKRSEAEKLTLVTRVQQLQGLLQNQSLQLQEQEKLLTKKDQALPV. | |
Affinity purified | |
RUO | |
80342 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction