Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAF3IP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TRAF3IP3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TRAF3IP3 Polyclonal specifically detects TRAF3IP3 in Human samples. It is validated for Western Blot.Specifications
TRAF3IP3 | |
Polyclonal | |
Rabbit | |
Q9Y228 | |
80342 | |
Synthetic peptides corresponding to TRAF3IP3(TRAF3 interacting protein 3) The peptide sequence was selected from the C terminal of TRAF3IP3. Peptide sequence DLQDQLKRSEAEKLTLVTRVQQLQGLLQNQSLQLQEQEKLLTKKDQALPV. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DJ434O14.3, FLJ44151, MGC117354, MGC163289, T3JAMTRAF3-interacting protein 3, TRAF3 interacting Jun N terminal kinase (JNK) activating modulator, TRAF3 interacting protein 3, TRAF3-interacting JNK-activating modulator | |
TRAF3IP3 | |
IgG | |
61 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title