Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAIL/TNFSF10 Antibody (2E5), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP140925AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP140925AF 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP140925AF Supplier Novus Biologicals Supplier No. NBP140925AF405

Mouse Monoclonal Antibody

TRAIL/TNFSF10 Monoclonal specifically detects TRAIL/TNFSF10 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen TRAIL/TNFSF10
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 200000
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10
Gene Symbols TNFSF10
Host Species Mouse
Immunogen Recombinant human CD253 (TRAIL) aa 95-281.
Purification Method Protein A purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8743
Test Specificity This antibody recognises the 281 amino acid protein termed TNF related apoptosis-inducing ligand (TRAIL), also known as CD253.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.