Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAIL/TNFSF10 Antibody (B-T24), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB388241 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388241 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388241 Supplier Novus Biologicals Supplier No. NBP314605IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAIL/TNFSF10 Monoclonal antibody specifically detects TRAIL/TNFSF10 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen TRAIL/TNFSF10
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone B-T24
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10
Host Species Mouse
Immunogen Recombinant human TRAIL/TNFSF10
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8743
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.