Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAIL/TNFSF10 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500220 0.1 mg
NB018185 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500220 Supplier Novus Biologicals Supplier No. NB500220
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

TRAIL/TNFSF10 Polyclonal specifically detects TRAIL/TNFSF10 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen TRAIL/TNFSF10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10
Gene Symbols TNFSF10
Host Species Rabbit
Immunogen residues [SNTLSSPNSKNE] of the N terminus of the TRAIL protein.
Molecular Weight of Antigen 36 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8743
Test Specificity Reacts with residues [SNTLSSPNSKNE] of the N terminus of the TRAIL protein.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.