Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAIL/TNFSF10 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.50 - $426.00
Specifications
| Antigen | TRAIL/TNFSF10 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
TRAIL/TNFSF10 Polyclonal specifically detects TRAIL/TNFSF10 in Human, Mouse samples. It is validated for Western Blot.Specifications
| TRAIL/TNFSF10 | |
| Western Blot 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Signal Transduction, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| 8743 | |
| residues [SNTLSSPNSKNE] of the N terminus of the TRAIL protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 36 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10 | |
| TNFSF10 | |
| IgG | |
| Affinity Purified | |
| Reacts with residues [SNTLSSPNSKNE] of the N terminus of the TRAIL protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title