Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR1/TNFRSF10A Antibody (32A1380), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP227350Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227350Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227350Y Supplier Novus Biologicals Supplier No. NBP227350AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen TRAILR1/TNFRSF10A
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 32A1380
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry
Gene Alias APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a
Gene Symbols TNFRSF10A
Host Species Mouse
Immunogen This monoclonal antibody was raised against a peptide corresponding to amino acid 2 to 21 of human DR4.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8797
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.