Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR1/TNFRSF10A Antibody (32A242), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB10056528Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056528Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056528Y Supplier Novus Biologicals Supplier No. NB10056528AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human, Mouse, Rat samples. It is validated for Immunoblot, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen TRAILR1/TNFRSF10A
Applications Immunoblot, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 32A242
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O00220
Gene Alias APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a
Gene Symbols TNFRSF10A
Host Species Mouse
Immunogen This monoclonal antibody was raised against a peptide corresponding to amino acid 2 to 21 of human DR4.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8797
Test Specificity In HeLa, a 57 kDa band should be observed.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.