Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR1/TNFRSF10A Antibody (DR-4-02), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP231343A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP231343A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP231343A70 Supplier Novus Biologicals Supplier No. NBP231343AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TRAILR1/TNFRSF10A
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone DR-4-02
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a
Gene Symbols TNFRSF10A
Host Species Mouse
Immunogen Fusion protein (Human). Fusion protein containing the extracellular part of DR4 and the constant part of the heavy chain of the human IgG1 (NM_003844.3).
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8797.0
Test Specificity The mouse monoclonal antibody DR-4-02 recognizes an extracellular epitope of TRAIL-R1 (DR4), a human death receptor 4 expressed in most human tissues (spleen, peripheral blood leucocytes, thymus) and in a variety of tumour-derived cell lines.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.