Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR2/TNFRSF10B Antibody (102), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB067175 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB067175 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB067175 Supplier Novus Biologicals Supplier No. NBP289513JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TRAILR2/TNFRSF10B Monoclonal specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen TRAILR2/TNFRSF10B
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 102
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TRAILR2/TNFRSF10B extracellular domain (Met 1-Glu 182; NP_003833.3).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8795
Test Specificity No cross-reactivity in ELISA with: Human TNFRSF10A/ CD261/ APO2 Human TNFRSF10D/ CD264/ DCR2/ TRAILR4
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.