Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR2/TNFRSF10B Antibody (131), Alexa Fluor™ 647
SDP

Catalog No. NB075183 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075183 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075183 Supplier Novus Biologicals Supplier No. NBP290575AF647

Rabbit Monoclonal Antibody

TRAILR2/TNFRSF10B Monoclonal specifically detects TRAILR2/TNFRSF10B in Mouse samples. It is validated for ELISA.

Specifications

Antigen TRAILR2/TNFRSF10B
Applications ELISA
Classification Monoclonal
Clone 131
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TRAILR2/TNFRSF10B (NP_064671.2; Met 1-Ser 177).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8795
Test Specificity No cross-reactivity in ELISA with: Human TNFRSF10B
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.