Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR2/TNFRSF10B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 41 publication
$192.47 - $517.59
Specifications
| Antigen | TRAILR2/TNFRSF10B |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 5 ug/ml, Flow Cytometry 1:20-1:2000, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 27740879), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Georgakis et al (2005)), Immunohistochemistry-Paraffin reported in scientific literature (Younes et al (2006)), Flow (Cell Surface) reported in scientific literature (KOYAMA (2003); PMID 16426839) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056618T
![]() |
Novus Biologicals
NB10056618SS |
0.025 mg |
Each for $192.47
|
|
|||||
NB10056618
![]() |
Novus Biologicals
NB10056618 |
0.1 mg |
Each for $517.59
|
|
|||||
Description
TRAILR2/TNFRSF10B Polyclonal specifically detects TRAILR2/TNFRSF10B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface).Specifications
| TRAILR2/TNFRSF10B | |
| Western Blot 5 ug/ml, Flow Cytometry 1:20-1:2000, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 27740879), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Georgakis et al (2005)), Immunohistochemistry-Paraffin reported in scientific literature (Younes et al (2006)), Flow (Cell Surface) reported in scientific literature (KOYAMA (2003); PMID 16426839) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 | |
| TNFRSF10B | |
| IgG | |
| Protein G purified |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Death Receptor Signaling Pathway | |
| PBS with 0.05% Sodium Azide | |
| 8795 | |
| Rabbit anti-DR5 polyclonal antibody was raised against a peptide corresponding to amino acids 388 to 407 (isoform 2) of human DR5 precursor (1,2). The same sequence is found in isoform 1 at amino acids 417-436. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title