Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR2/TNFRSF10B Antibody, Novus Biologicals™
SDP

Catalog No. NB10056618T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056618T 0.025 mg
NB10056618 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056618T Supplier Novus Biologicals Supplier No. NB10056618SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 41 publication

TRAILR2/TNFRSF10B Polyclonal specifically detects TRAILR2/TNFRSF10B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface).

Specifications

Antigen TRAILR2/TNFRSF10B
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 5 ug/ml, Flow Cytometry 1:20-1:2000, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 27740879), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Georgakis et al (2005)), Immunohistochemistry-Paraffin reported in scientific literature (Younes et al (2006)), Flow (Cell Surface) reported in scientific literature (KOYAMA (2003); PMID 16426839)
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9
Gene Symbols TNFRSF10B
Host Species Rabbit
Immunogen Rabbit anti-DR5 polyclonal antibody was raised against a peptide corresponding to amino acids 388 to 407 (isoform 2) of human DR5 precursor (1,2). The same sequence is found in isoform 1 at amino acids 417-436.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Death Receptor Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8795
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.