Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR2/TNFRSF10B Antibody (HS201), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB172816 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172816 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172816 Supplier Novus Biologicals Supplier No. NBP280067DL594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRAILR2/TNFRSF10B Monoclonal antibody specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence

Specifications

Antigen TRAILR2/TNFRSF10B
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone HS201
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9
Host Species Mouse
Immunogen Recombinant human TRAILR2/TNFRSF10B (DR5).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8795
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.