Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR2/TNFRSF10B Antibody (OTI2A5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP274578 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274578 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274578 Supplier Novus Biologicals Supplier No. NBP274578
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAILR2/TNFRSF10B Monoclonal specifically detects TRAILR2/TNFRSF10B in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TRAILR2/TNFRSF10B
Applications Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Monoclonal
Clone OTI2A5
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-500, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9
Gene Symbols TNFRSF10B
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 203-411 of human TNFRSF10B(NP_671716) produced in E.coli.
Molecular Weight of Antigen 39.4 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8795
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.