Learn More
Description
Specifications
Specifications
| Antigen | TRAILR2/TNFRSF10B |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 102 |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 |
| Host Species | Rabbit |
| Immunogen | Recombinant Human TRAILR2/TNFRSF10B protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
