Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR3/TNFRSF10C Antibody (TR3.06), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB172922 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172922 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172922 Supplier Novus Biologicals Supplier No. NBP280071R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRAILR3/TNFRSF10C Monoclonal antibody specifically detects TRAILR3/TNFRSF10C in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TRAILR3/TNFRSF10C
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TR3.06
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Antagonist decoy receptor for TRAIL/Apo-2L, CD263, CD263 antigen, cytotoxic TRAIL receptor-3, DcR1, DCR1MGC149502, Decoy receptor 1, Decoy TRAIL receptor without death domain, LITMGC149501, Lymphocyte inhibitor of TRAIL, TNFRSF10C, TRAIL receptor 3, TRAIL receptor without an intracellular domain, TRAIL-R3, TRAILR3TNF-related apoptosis-inducing ligand receptor 3, TRIDDCR1-TNFR, tumor necrosis factor receptor superfamily member 10C, tumor necrosis factor receptor superfamily, member 10c, decoy without anintracellular domain
Host Species Mouse
Immunogen Recombinant human TRAILR3/TNFRSF10C (DcR1).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cell Biology, Chromatin Research, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8794
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.