Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAILR4/TNFRSF10D/DcR2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB214858 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB214858 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB214858 Supplier Novus Biologicals Supplier No. NBP176985AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRAILR4/TNFRSF10D/DcR2 Polyclonal antibody specifically detects TRAILR4/TNFRSF10D/DcR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Cell Surface)

Specifications

Antigen TRAILR4/TNFRSF10D/DcR2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid peptide near the center of human DcR2. The immunogen is located within amino acids 230 - 280 of DcR2. Amino Acid Squence: GGPERVHRVLFRRRS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8793
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.