Learn More
TRAILR4/TNFRSF10D/DcR2 Rabbit anti-Human, Clone: 43, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | TRAILR4/TNFRSF10D/DcR2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 43 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain |
| Host Species | Rabbit |
| Immunogen | Recombinant Human TRAILR4/TNFRSF10D/DcR2 protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.