Learn More
Description
Specifications
Specifications
| Antigen | TRAIP |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Alias | RNF206TRIPRING finger protein 206, TRAF interacting protein, TRAF-interacting protein |
| Gene Symbols | TRAIP |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acids 294-313 (ESPAPVEVNLKLRRPSFRDD) of human TRIP was used as immunogen; GenBank no. gi 40807469 ref NP_005870.2 . |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
