Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
TRAM 39
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                5 mg
                            
                        
                            
                                25 mg
                            
                        
                    Description
TRAM 39 is a potent intermediate conductance Ca2+-activated K+ channel (KCa3.1) blocker (Kd = 60 nM). Has no effect on cytochrome p450 activity. Inhibits I-EBIO -stimulated increases in rat artery membrane potential ex vivo. Also diminishes LPS-induced cryptidin (mammalian α-defensin) release from paneth cells in vitro.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
                
            Specifications
Specifications
| Chemical Name or Material | 2-Chloro-α,α-diphenylbenzeneacetonitrile | 
| CAS | 197525-99-8 | 
| Assay Percent Range | >99% | 
| Quantity | 25 mg | 
| Molecular Formula | C20H14ClN | 
| Formula Weight | Observed MW: 303.78 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                