Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAM/TICAM2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224638SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224638SS 0.025 mg
NBP224638 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224638SS Supplier Novus Biologicals Supplier No. NBP224638SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRAM/TICAM2 Polyclonal specifically detects TRAM/TICAM2 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TRAM/TICAM2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q86XR7
Gene Alias MGC129876, MGC129877, Putative NF-kappa-B-activating protein 502, TICAM-2TIR domain-containing adapter molecule 2, TIRAP3, TIRPcytoplasmic adaptor, toll/interleukin-1 receptor (TIR) domain-containing adapter protein, Toll/interleukin-1 receptor domain-containing protein, toll-like receptor adaptor molecule 2, Toll-like receptor adaptor protein 3, TRAMNF-kappa-B-activating protein 502, TRIF-related adapter molecule, TRIF-related adaptor molecule
Gene Symbols TICAM2
Host Species Rabbit
Immunogen Within the range of amino acids 87-124 of human TRAM (TICAM2) protein were used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 353376
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, and rhesus monkey, and 93% homologous in rat and mouse.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.