Learn More
Description
Specifications
Specifications
| Antigen | TRAM/TICAM2 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Formulation | 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Alias | MGC129876, MGC129877, Putative NF-kappa-B-activating protein 502, TICAM-2TIR domain-containing adapter molecule 2, TIRAP3, TIRPcytoplasmic adaptor, toll/interleukin-1 receptor (TIR) domain-containing adapter protein, Toll/interleukin-1 receptor domain-containing protein, toll-like receptor adaptor molecule 2, Toll-like receptor adaptor protein 3, TRAMNF-kappa-B-activating protein 502, TRIF-related adapter molecule, TRIF-related adaptor molecule |
| Gene Symbols | TICAM2 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
