Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAM/TICAM2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen TRAM/TICAM2
Concentration 0.5 mg/mL
Dilution Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224686SS
SDP
View Documents
Novus Biologicals
NBP224686SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224686
SDP
View Documents
Novus Biologicals
NBP224686
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

TRAM/TICAM2 Polyclonal specifically detects TRAM/TICAM2 in Human, Mouse, Rat, Bovine, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TRAM/TICAM2
Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Rat, Bovine, Opossum, Primate
MGC129876, MGC129877, Putative NF-kappa-B-activating protein 502, TICAM-2TIR domain-containing adapter molecule 2, TIRAP3, TIRPcytoplasmic adaptor, toll/interleukin-1 receptor (TIR) domain-containing adapter protein, Toll/interleukin-1 receptor domain-containing protein, toll-like receptor adaptor molecule 2, Toll-like receptor adaptor protein 3, TRAMNF-kappa-B-activating protein 502, TRIF-related adapter molecule, TRIF-related adaptor molecule
TICAM2
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, wild boar, horse, mouse, chimpanzee, chicken, dog, rat and macaque and 94% homologous in pufferfish.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Immunology
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
353376
A portion of amino acids 150-200 of TAG/TIRAP3b was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.