Learn More
TRANCE/TNFSF11/RANK L Antibody (12A380), Alexa Fluor™ 532, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | TRANCE/TNFSF11/RANK L |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 12A380 |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Flow (Cell Surface) |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 |
| Gene Symbols | TNFSF11 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.