Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRANCE/TNFSF11/RANK L Antibody (12A380), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP227402UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227402UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227402UV Supplier Novus Biologicals Supplier No. NBP227402UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Flow (Cell Surface).

Specifications

Antigen TRANCE/TNFSF11/RANK L
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 12A380
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Frozen, Flow (Cell Surface)
Gene Alias CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11
Gene Symbols TNFSF11
Host Species Mouse
Immunogen A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE was used as immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8600
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.